Product Details

SNP ID
rs150002191
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:113799303 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATGACAGCTGCAAATCGGCTACA[C/T]AGGTCATAGGGAGAATTGGGTCGAA
Phenotype
MIM: 610748
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC390251 PubMed Links

Gene Details

Gene
LOC390251
Gene Name
SH3 domain containing GRB2 like 1 pseudogene
There are no transcripts associated with this gene.

Gene
USP28
Gene Name
ubiquitin specific peptidase 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301029.1 3425 Silent Mutation NP_001287958.1
NM_020886.3 3425 Silent Mutation NP_065937.1
XM_005271630.2 3425 Silent Mutation XP_005271687.1
XM_005271631.2 3425 Silent Mutation XP_005271688.1
XM_005271632.2 3425 Silent Mutation XP_005271689.1
XM_005271633.4 3425 Silent Mutation XP_005271690.1
XM_005271636.2 3425 Silent Mutation XP_005271693.1
XM_005271637.2 3425 Silent Mutation XP_005271694.1
XM_005271638.2 3425 Silent Mutation XP_005271695.1
XM_005271639.2 3425 Silent Mutation XP_005271696.1
XM_011542936.2 3425 Silent Mutation XP_011541238.1
XM_011542938.1 3425 Silent Mutation XP_011541240.1
XM_011542941.1 3425 Silent Mutation XP_011541243.1
XM_011542942.1 3425 Silent Mutation XP_011541244.1
XM_017018056.1 3425 Silent Mutation XP_016873545.1
XM_017018057.1 3425 Silent Mutation XP_016873546.1
XM_017018058.1 3425 Silent Mutation XP_016873547.1
XM_017018059.1 3425 Silent Mutation XP_016873548.1
XM_017018060.1 3425 Silent Mutation XP_016873549.1
XM_017018061.1 3425 Silent Mutation XP_016873550.1
XM_017018062.1 3425 Silent Mutation XP_016873551.1
XM_017018063.1 3425 Silent Mutation XP_016873552.1
XM_017018064.1 3425 Silent Mutation XP_016873553.1
XM_017018065.1 3425 Silent Mutation XP_016873554.1
XM_017018066.1 3425 Silent Mutation XP_016873555.1
XM_017018067.1 3425 Silent Mutation XP_016873556.1
XM_017018068.1 3425 Silent Mutation XP_016873557.1
XM_017018069.1 3425 Intron XP_016873558.1

View Full Product Details