Product Details

SNP ID
rs150075556
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:290888 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGCTGCAGGCCAGGGGAGCTCT[A/G]TATACGGCTCACGCACAGGCCTGGG
Phenotype
MIM: 614757 MIM: 609650 MIM: 617032
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
IFITM5 PubMed Links
Additional Information
For this assay, SNP(s) [rs72636977] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
IFITM5
Gene Name
interferon induced transmembrane protein 5
There are no transcripts associated with this gene.

Gene
NLRP6
Gene Name
NLR family pyrin domain containing 6
There are no transcripts associated with this gene.

Gene
PGGHG
Gene Name
protein-glucosylgalactosylhydroxylysine glucosidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025092.4 1833 Silent Mutation CTA,CTG L227L NP_079368.3
XM_011520383.2 1833 Silent Mutation CTA,CTG L227L XP_011518685.1
XM_011520384.2 1833 Silent Mutation CTA,CTG L227L XP_011518686.1
XM_011520385.2 1833 Silent Mutation CTA,CTG L227L XP_011518687.1
XM_011520386.2 1833 Silent Mutation CTA,CTG L227L XP_011518688.1
XM_011520387.2 1833 Silent Mutation CTA,CTG L227L XP_011518689.1
XM_017018355.1 1833 Silent Mutation CTA,CTG L227L XP_016873844.1

View Full Product Details