Product Details

SNP ID
rs150226588
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:31444305 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACATTTGCCAGTGTGAATTATGTC[C/T]ATTGGGCTTTCTTTGTCCCTGGTTT
Phenotype
MIM: 612323
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IMMP1L PubMed Links

Gene Details

Gene
IMMP1L
Gene Name
inner mitochondrial membrane peptidase subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304274.1 Intron NP_001291203.1
NM_144981.2 Intron NP_659418.1
XM_005252812.3 Intron XP_005252869.1
XM_011519942.1 Intron XP_011518244.1
XM_011519943.2 Intron XP_011518245.1
XM_011519944.2 Intron XP_011518246.1
XM_011519945.2 Intron XP_011518247.1
XM_011519946.2 Intron XP_011518248.1
XM_011519947.2 Intron XP_011518249.1
XM_017017305.1 Intron XP_016872794.1
XM_017017306.1 Intron XP_016872795.1
XM_017017307.1 Intron XP_016872796.1

View Full Product Details