Product Details

SNP ID
rs150533004
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:75587310 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGATTCTTTGGAGGCTGTAGGA[C/T]TGTGGGGCCTTGATCCTTAACTAGT
Phenotype
MIM: 601783
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
MAP6 PubMed Links

Gene Details

Gene
MAP6
Gene Name
microtubule associated protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033063.1 4843 Missense Mutation ATC,GTC I731V NP_149052.1
NM_207577.1 4843 Intron NP_997460.1
XM_006718556.3 4843 Intron XP_006718619.1
XM_011545039.2 4843 Intron XP_011543341.1
XM_017017755.1 4843 Missense Mutation ATC,GTC I747V XP_016873244.1
XM_017017756.1 4843 Missense Mutation ATC,GTC I446V XP_016873245.1
XM_017017757.1 4843 Missense Mutation ATC,GTC I430V XP_016873246.1
XM_017017758.1 4843 Missense Mutation ATC,GTC I418V XP_016873247.1

View Full Product Details