Product Details

SNP ID
rs150681380
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65340445 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGCCACAATTACAATGCTCGCCT[C/T]TGTGCTGAGCGCAGCGTGCGCCTGC
Phenotype
MIM: 601671
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DPF2 PubMed Links

Gene Details

Gene
DPF2
Gene Name
double PHD fingers 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006268.4 Intron NP_006259.1
XM_005274149.2 Intron XP_005274206.1
XM_017018101.1 Intron XP_016873590.1
XM_017018102.1 Intron XP_016873591.1

View Full Product Details