Product Details

SNP ID
rs150718407
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:46679465 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTGGCACCCTCTGGCTTTCATCA[A/G]TGTCTATGAGGAAAGGAGGCCCGGG
Phenotype
MIM: 602732 MIM: 615088
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP1 PubMed Links

Gene Details

Gene
ARHGAP1
Gene Name
Rho GTPase activating protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004308.3 948 Missense Mutation ACT,ATT T344I NP_004299.1
XM_011520095.1 948 Missense Mutation ACT,ATT T344I XP_011518397.1
XM_017017720.1 948 Missense Mutation ACT,ATT T300I XP_016873209.1
Gene
ATG13
Gene Name
autophagy related 13
There are no transcripts associated with this gene.

View Full Product Details