Product Details

SNP ID
rs151161255
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:209306 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCACGTTTGATGATGCCCAACA[A/G]GAGGACCGGAAGGTGGGTGCTGGCC
Phenotype
MIM: 615417 MIM: 608356 MIM: 609146 MIM: 604481
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BET1L PubMed Links

Gene Details

Gene
BET1L
Gene Name
Bet1 golgi vesicular membrane trafficking protein like
There are no transcripts associated with this gene.

Gene
MIR6743
Gene Name
microRNA 6743
There are no transcripts associated with this gene.

Gene
ODF3
Gene Name
outer dense fiber of sperm tails 3
There are no transcripts associated with this gene.

Gene
RIC8A
Gene Name
RIC8 guanine nucleotide exchange factor A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286134.1 650 Silent Mutation CAA,CAG Q40Q NP_001273063.1
NM_021932.5 650 Silent Mutation CAA,CAG Q40Q NP_068751.4
XM_005253052.3 650 Silent Mutation CAA,CAG Q44Q XP_005253109.1
XM_017018107.1 650 UTR 5 XP_016873596.1
Gene
SIRT3
Gene Name
sirtuin 3
There are no transcripts associated with this gene.

View Full Product Details