Product Details
- SNP ID
-
rs151195047
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:61102614 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCACCTTGTACCTGCTGGGGATGCT[A/G]GGTGAGTACCCCTCCCAGGTGTCCT
- Phenotype
-
MIM: 153340
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CD5
PubMed Links
Gene Details
- Gene
- CD5
- Gene Name
- CD5 molecule
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014207.3 |
157 |
Silent Mutation |
CTA,CTG |
L18L |
NP_055022.2 |
View Full Product Details