Product Details

SNP ID
rs138152846
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:42328641 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTTAGTGCAGTACATTCGGGCTT[A/G]TAAAAGAAGTATCCTTAAAGCGTTG
Phenotype
MIM: 608150 MIM: 610750
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR7851 PubMed Links

Gene Details

Gene
MIR7851
Gene Name
microRNA 7851
There are no transcripts associated with this gene.

Gene
PPHLN1
Gene Name
periphilin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143787.1 Intron NP_001137259.1
NM_001143788.1 Intron NP_001137260.1
NM_001143789.1 Intron NP_001137261.1
NM_016488.6 Intron NP_057572.5
NM_201438.1 Intron NP_958846.1
NM_201439.1 Intron NP_958847.1
NM_201440.1 Intron NP_958848.1
NM_201515.1 Intron NP_958923.1
XM_005268962.3 Intron XP_005269019.1
XM_011538459.2 Intron XP_011536761.1
XM_011538465.2 Intron XP_011536767.1
XM_011538467.2 Intron XP_011536769.1
XM_011538470.2 Intron XP_011536772.1
XM_011538471.2 Intron XP_011536773.1
XM_017019434.1 Intron XP_016874923.1
XM_017019435.1 Intron XP_016874924.1
XM_017019436.1 Intron XP_016874925.1
XM_017019437.1 Intron XP_016874926.1
XM_017019438.1 Intron XP_016874927.1
XM_017019439.1 Intron XP_016874928.1
XM_017019440.1 Intron XP_016874929.1
XM_017019441.1 Intron XP_016874930.1
XM_017019442.1 Intron XP_016874931.1
XM_017019443.1 Intron XP_016874932.1
XM_017019444.1 Intron XP_016874933.1
XM_017019445.1 Intron XP_016874934.1
XM_017019446.1 Intron XP_016874935.1
XM_017019447.1 Intron XP_016874936.1
XM_017019448.1 Intron XP_016874937.1
XM_017019449.1 Intron XP_016874938.1
XM_017019450.1 Intron XP_016874939.1
XM_017019451.1 Intron XP_016874940.1
XM_017019452.1 Intron XP_016874941.1
XM_017019453.1 Intron XP_016874942.1
Gene
ZCRB1
Gene Name
zinc finger CCHC-type and RNA binding motif containing 1
There are no transcripts associated with this gene.

View Full Product Details