Product Details

SNP ID
rs138450211
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:12813380 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCTCACACAAGATGGCGGCACCC[A/G]AGGAACACGATTCTCCGACCGAAGC
Phenotype
MIM: 615428
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDX47 PubMed Links

Gene Details

Gene
DDX47
Gene Name
DEAD-box helicase 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016355.3 35 Missense Mutation AAG,GAG K5E NP_057439.2
NM_201224.1 35 Missense Mutation AAG,GAG K5E NP_957518.1

View Full Product Details