Product Details
- SNP ID
-
rs139528592
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:124913733 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTCACTGGGCTCAACCTCGAGGGT[A/G]ATGGTCTTACCAGTCAGAGTCTTCA
- Phenotype
-
MIM: 191340
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR5188
PubMed Links
Gene Details
- Gene
- MIR5188
- Gene Name
- microRNA 5188
There are no transcripts associated with this gene.
- Gene
- UBC
- Gene Name
- ubiquitin C
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021009.6 |
497 |
Silent Mutation |
ATC,ATT |
I13I |
NP_066289.3 |
View Full Product Details