Product Details

SNP ID
rs140174092
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56344111 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCTGGCTCTGGCTTAAGCTCCAG[C/T]GGTTGTTGCAGTTCATCCACCTGTC
Phenotype
MIM: 605580 MIM: 600556
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL23A PubMed Links

Gene Details

Gene
IL23A
Gene Name
interleukin 23 subunit alpha
There are no transcripts associated with this gene.

Gene
STAT2
Gene Name
signal transducer and activator of transcription 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005419.3 1660 Silent Mutation CCA,CCG P709P NP_005410.1
NM_198332.1 1660 Silent Mutation CCA,CCG P705P NP_938146.1
XM_005269111.3 1660 Intron XP_005269168.2
XM_011538697.2 1660 Silent Mutation CCA,CCG P717P XP_011536999.1
XM_011538698.2 1660 Silent Mutation CCA,CCG P713P XP_011537000.1
XM_011538699.2 1660 Intron XP_011537001.1
XM_011538700.2 1660 Silent Mutation CCA,CCG P473P XP_011537002.1
XM_017019904.1 1660 Silent Mutation CCA,CCG P465P XP_016875393.1

View Full Product Details