Product Details

SNP ID
rs140329323
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:106997370 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCATTCCCATTAGGATTAGAAGG[C/T]ACTGATGCCAGAAGACCTAAAGGAC
Phenotype
MIM: 601933
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRY1 PubMed Links

Gene Details

Gene
CRY1
Gene Name
cryptochrome circadian clock 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004075.4 2405 Silent Mutation GTA,GTG V503V NP_004066.1
XM_017018832.1 2405 Silent Mutation GTA,GTG V475V XP_016874321.1

View Full Product Details