Product Details

SNP ID
rs141733387
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:1971401 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATGCCGTTCTTTGGAAATTCTCA[A/G]TTGAAAAAAAGTGATGTTTGAGGCA
Phenotype
MIM: 114205 MIM: 609843
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CACNA1C PubMed Links

Gene Details

Gene
CACNA1C
Gene Name
calcium voltage-gated channel subunit alpha1 C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000719.6 Intron NP_000710.5
NM_001129827.1 Intron NP_001123299.1
NM_001129829.1 Intron NP_001123301.1
NM_001129830.2 Intron NP_001123302.2
NM_001129831.1 Intron NP_001123303.1
NM_001129832.1 Intron NP_001123304.1
NM_001129833.1 Intron NP_001123305.1
NM_001129834.1 Intron NP_001123306.1
NM_001129835.1 Intron NP_001123307.1
NM_001129836.1 Intron NP_001123308.1
NM_001129837.1 Intron NP_001123309.1
NM_001129838.1 Intron NP_001123310.1
NM_001129839.1 Intron NP_001123311.1
NM_001129840.1 Intron NP_001123312.1
NM_001129841.1 Intron NP_001123313.1
NM_001129842.1 Intron NP_001123314.1
NM_001129843.1 Intron NP_001123315.1
NM_001129844.1 Intron NP_001123316.1
NM_001129846.1 Intron NP_001123318.1
NM_001167623.1 Intron NP_001161095.1
NM_001167624.2 Intron NP_001161096.2
NM_001167625.1 Intron NP_001161097.1
NM_199460.3 Intron NP_955630.3
XM_006719017.2 Intron XP_006719080.1
XM_011521020.2 Intron XP_011519322.1
XM_011521023.2 Intron XP_011519325.1
XM_017019926.1 Intron XP_016875415.1
XM_017019927.1 Intron XP_016875416.1
XM_017019928.1 Intron XP_016875417.1
XM_017019929.1 Intron XP_016875418.1
XM_017019930.1 Intron XP_016875419.1
XM_017019931.1 Intron XP_016875420.1
XM_017019932.1 Intron XP_016875421.1
XM_017019933.1 Intron XP_016875422.1
XM_017019934.1 Intron XP_016875423.1
XM_017019935.1 Intron XP_016875424.1
XM_017019936.1 Intron XP_016875425.1
XM_017019937.1 Intron XP_016875426.1
XM_017019938.1 Intron XP_016875427.1
XM_017019939.1 Intron XP_016875428.1
XM_017019940.1 Intron XP_016875429.1
XM_017019941.1 Intron XP_016875430.1
XM_017019942.1 Intron XP_016875431.1
XM_017019943.1 Intron XP_016875432.1
XM_017019944.1 Intron XP_016875433.1
XM_017019945.1 Intron XP_016875434.1
XM_017019946.1 Intron XP_016875435.1
XM_017019947.1 Intron XP_016875436.1
XM_017019948.1 Intron XP_016875437.1
XM_017019949.1 Intron XP_016875438.1
XM_017019950.1 Intron XP_016875439.1
XM_017019951.1 Intron XP_016875440.1
XM_017019952.1 Intron XP_016875441.1
XM_017019953.1 Intron XP_016875442.1
XM_017019954.1 Intron XP_016875443.1
XM_017019955.1 Intron XP_016875444.1
Gene
DCP1B
Gene Name
decapping mRNA 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319292.1 Intron NP_001306221.1
NM_152640.4 Intron NP_689853.3
XM_011520927.2 Intron XP_011519229.1

View Full Product Details