Product Details

SNP ID
rs141901432
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:106065528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGTTTAAAACCATGACATAGAA[C/T]ACTTAGATTTGGGGGAAAAAAAGCA
Phenotype
MIM: 608130
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUAK1 PubMed Links

Gene Details

Gene
NUAK1
Gene Name
NUAK family kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014840.2 4640 UTR 3 NP_055655.1

View Full Product Details