Product Details

SNP ID
rs141968055
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:85280408 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGCAAAGCGTCTGCAGGCAAATG[C/T]GTGCAGGCCTTCGGACCCCTGCCCC
Phenotype
MIM: 601527
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALX1 PubMed Links

Gene Details

Gene
ALX1
Gene Name
ALX homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006982.2 151 Silent Mutation TGC,TGT C49C NP_008913.2
XM_011538782.2 151 Intron XP_011537084.1
Gene
LRRIQ1
Gene Name
leucine rich repeats and IQ motif containing 1
There are no transcripts associated with this gene.

View Full Product Details