Product Details

SNP ID
rs142229500
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48569136 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTGAGGGACCTGGCTGCTCTTGC[A/G]CCAAAGCTTATTACTGATCTGGAAG
Phenotype
MIM: 149750
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LALBA PubMed Links

Gene Details

Gene
LALBA
Gene Name
lactalbumin alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002289.2 342 Missense Mutation CGC,TGC R80C NP_002280.1
XM_006719395.3 342 Missense Mutation CGC,TGC R87C XP_006719458.1

View Full Product Details