Product Details

SNP ID
rs142551471
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56343229 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTCCCCTCTGTACCCATGTTATG[C/G]TTTCACCTCTCACCCCAATGGAGTC
Phenotype
MIM: 605580 MIM: 600556
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IL23A PubMed Links

Gene Details

Gene
IL23A
Gene Name
interleukin 23 subunit alpha
There are no transcripts associated with this gene.

Gene
PAN2
Gene Name
PAN2 poly(A) specific ribonuclease subunit
There are no transcripts associated with this gene.

Gene
STAT2
Gene Name
signal transducer and activator of transcription 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005419.3 2249 UTR 3 NP_005410.1
NM_198332.1 2249 UTR 3 NP_938146.1
XM_005269111.3 2249 Intron XP_005269168.2
XM_011538697.2 2249 UTR 3 XP_011536999.1
XM_011538698.2 2249 UTR 3 XP_011537000.1
XM_011538699.2 2249 Intron XP_011537001.1
XM_011538700.2 2249 UTR 3 XP_011537002.1
XM_017019904.1 2249 UTR 3 XP_016875393.1

View Full Product Details