Product Details

SNP ID
rs143287957
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:7133133 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTGCCTTGGACAAGGATGCCCC[A/G]CTGCGCTATGCAGGTAATTGGGATT
Phenotype
MIM: 611324 MIM: 611866
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CLSTN3 PubMed Links

Gene Details

Gene
CLSTN3
Gene Name
calsyntenin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014718.3 452 Silent Mutation CCA,CCG P58P NP_055533.2
XM_006719163.3 452 Silent Mutation CCA,CCG P70P XP_006719226.1
Gene
RBP5
Gene Name
retinol binding protein 5
There are no transcripts associated with this gene.

View Full Product Details