Product Details

SNP ID
rs143587024
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:71689600 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTATAATTTTTCAGGTAACTTAC[A/G]ACCTATTTCTCCGTGGCGTTGGCTG
Phenotype
MIM: 612531
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
THAP2 PubMed Links

Gene Details

Gene
THAP2
Gene Name
THAP domain containing 2
There are no transcripts associated with this gene.

Gene
TMEM19
Gene Name
transmembrane protein 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018279.3 723 Missense Mutation CAA,CGA Q47R NP_060749.2

View Full Product Details