Product Details

SNP ID
rs143590566
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:14770902 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCACGGATCACGTTCTCCAGGAA[A/G]ACTTTGAGGACTCCCCGGGTCTCCT
Phenotype
MIM: 615069
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C12orf60 PubMed Links

Gene Details

Gene
C12orf60
Gene Name
chromosome 12 open reading frame 60
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175874.3 230 Intron NP_787070.2
XM_005253322.4 230 Intron XP_005253379.1
XM_011520568.2 230 Intron XP_011518870.1
XM_011520569.2 230 Intron XP_011518871.1
XM_017018871.1 230 Intron XP_016874360.1
XM_017018872.1 230 Intron XP_016874361.1
XM_017018873.1 230 Intron XP_016874362.1
XM_017018874.1 230 Intron XP_016874363.1
Gene
H2AFJ
Gene Name
H2A histone family member J
There are no transcripts associated with this gene.

Gene
HIST4H4
Gene Name
histone cluster 4, H4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175054.2 230 Silent Mutation GTC,GTT V61V NP_778224.1
Gene
LOC105369669
Gene Name
uncharacterized LOC105369669
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011520896.2 230 Intron XP_011519198.1

View Full Product Details