Product Details

SNP ID
rs144289367
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48858154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGACACAGCATGGATGCCGTCC[A/G]AAAGATGCTGTGGATGCTCTTTTCT
Phenotype
MIM: 612172 MIM: 609038
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDX23 PubMed Links

Gene Details

Gene
DDX23
Gene Name
DEAD-box helicase 23
There are no transcripts associated with this gene.

Gene
RND1
Gene Name
Rho family GTPase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014470.3 644 Missense Mutation CGG,TGG R181W NP_055285.1

View Full Product Details