Product Details

SNP ID
rs144662472
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48861094 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTTGCAGCCAATGAGCAAAACG[C/G]GGGTGCTGGGACAATAATCTAGGAT
Phenotype
MIM: 612172 MIM: 609038
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DDX23 PubMed Links

Gene Details

Gene
DDX23
Gene Name
DEAD-box helicase 23
There are no transcripts associated with this gene.

Gene
RND1
Gene Name
Rho family GTPase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014470.3 459 Missense Mutation CCC,CGC P119R NP_055285.1

View Full Product Details