Product Details

SNP ID
rs144680004
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:51889174 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGGACAAAGGAGGGGACACGGC[C/G]CTCATGTTGGCTGCCCAAGCAGGTG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD33 PubMed Links

Gene Details

Gene
ANKRD33
Gene Name
ankyrin repeat domain 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130015.1 602 Silent Mutation GCC,GCG A33A NP_001123487.1
NM_001304459.1 602 Silent Mutation GCC,GCG A33A NP_001291388.1
NM_001304460.1 602 Intron NP_001291389.1
NM_182608.3 602 Silent Mutation GCC,GCG A168A NP_872414.3
XM_017019258.1 602 Silent Mutation GCC,GCG A177A XP_016874747.1

View Full Product Details