Product Details

SNP ID
rs145082582
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:66304958 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTAAAGAGTGTGAGGTCTCCAGT[A/G]ATGATGTTAATAAATTTTTAACATG
Phenotype
MIM: 614539
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HELB PubMed Links

Gene Details

Gene
HELB
Gene Name
DNA helicase B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033647.4 474 Missense Mutation AAT,GAT N139D NP_387467.2
XM_005269234.2 474 Missense Mutation AAT,GAT N139D XP_005269291.1
XM_017020230.1 474 Missense Mutation AAT,GAT N139D XP_016875719.1

View Full Product Details