Product Details

SNP ID
rs145372838
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:122172588 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAGTTTGTCGAGGTGCTCTATGA[A/T]CTCATCAATAACAGATTTGTTGTCT
Phenotype
MIM: 609509
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
IL31 PubMed Links

Gene Details

Gene
IL31
Gene Name
interleukin 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014336.1 346 Missense Mutation ATC,TTC I107F NP_001014358.1
Gene
LRRC43
Gene Name
leucine rich repeat containing 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098519.1 346 Intron NP_001091989.1
NM_152759.4 346 Intron NP_689972.3
XM_017019124.1 346 Intron XP_016874613.1
XM_017019125.1 346 Intron XP_016874614.1

View Full Product Details