Product Details

SNP ID
rs145530655
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:99648218 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACCGTATTACACGGCCCAAAGCA[A/G]CCCCGCAATGGGCATGTTTAACACC
Phenotype
MIM: 607815
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKS1B PubMed Links

Gene Details

Gene
ANKS1B
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204065.1 469 Intron NP_001190994.1
NM_001204066.1 469 Intron NP_001190995.1
NM_001204067.1 469 Intron NP_001190996.1
NM_001204068.1 469 Intron NP_001190997.1
NM_001204069.1 469 Intron NP_001190998.1
NM_001204070.1 469 Intron NP_001190999.1
NM_001204079.1 469 Intron NP_001191008.1
NM_001204080.1 469 Intron NP_001191009.1
NM_001204081.1 469 Intron NP_001191010.1
NM_020140.3 469 Intron NP_064525.1
NM_152788.4 469 Intron NP_690001.3
NM_181670.3 469 Intron NP_858056.2
XM_005269028.4 469 Intron XP_005269085.1
XM_005269029.4 469 Intron XP_005269086.1
XM_005269032.3 469 Intron XP_005269089.1
XM_006719504.3 469 Intron XP_006719567.1
XM_006719505.3 469 Intron XP_006719568.1
XM_006719506.3 469 Intron XP_006719569.1
XM_006719507.3 469 Intron XP_006719570.1
XM_006719508.3 469 Intron XP_006719571.1
XM_006719509.3 469 Intron XP_006719572.1
XM_006719510.3 469 Intron XP_006719573.1
XM_006719512.3 469 Intron XP_006719575.1
XM_006719513.3 469 Intron XP_006719576.1
XM_006719514.3 469 Intron XP_006719577.1
XM_011538571.2 469 Intron XP_011536873.1
XM_017019651.1 469 Intron XP_016875140.1
XM_017019652.1 469 Intron XP_016875141.1
XM_017019653.1 469 Intron XP_016875142.1
XM_017019654.1 469 Intron XP_016875143.1
XM_017019655.1 469 Intron XP_016875144.1
XM_017019656.1 469 Intron XP_016875145.1
XM_017019657.1 469 Intron XP_016875146.1
XM_017019658.1 469 Intron XP_016875147.1
XM_017019659.1 469 Intron XP_016875148.1
XM_017019660.1 469 Intron XP_016875149.1
XM_017019661.1 469 Intron XP_016875150.1
XM_017019662.1 469 Intron XP_016875151.1
XM_017019663.1 469 Intron XP_016875152.1
XM_017019664.1 469 Intron XP_016875153.1
XM_017019665.1 469 Intron XP_016875154.1
Gene
FAM71C
Gene Name
family with sequence similarity 71 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153364.3 469 Missense Mutation AAC,AGC N15S NP_699195.1

View Full Product Details