Product Details

SNP ID
rs146104637
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:42313192 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTTTTTTGTTCTTCTTCAATTT[A/T]GGCTTGCTGTGATTCAAAAAACAAA
Phenotype
MIM: 610750
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZCRB1 PubMed Links

Gene Details

Gene
ZCRB1
Gene Name
zinc finger CCHC-type and RNA binding motif containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033114.3 629 Nonsense Mutation AAA,TAA K177* NP_149105.3
XM_017020124.1 629 Nonsense Mutation AAA,TAA K136* XP_016875613.1

View Full Product Details