Product Details

SNP ID
rs146401531
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:99648206 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTGCTGTATGCTACCGTATTACA[C/T]GGCCCAAAGCAGCCCCGCAATGGGC
Phenotype
MIM: 607815
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKS1B PubMed Links

Gene Details

Gene
ANKS1B
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204065.1 457 Intron NP_001190994.1
NM_001204066.1 457 Intron NP_001190995.1
NM_001204067.1 457 Intron NP_001190996.1
NM_001204068.1 457 Intron NP_001190997.1
NM_001204069.1 457 Intron NP_001190998.1
NM_001204070.1 457 Intron NP_001190999.1
NM_001204079.1 457 Intron NP_001191008.1
NM_001204080.1 457 Intron NP_001191009.1
NM_001204081.1 457 Intron NP_001191010.1
NM_020140.3 457 Intron NP_064525.1
NM_152788.4 457 Intron NP_690001.3
NM_181670.3 457 Intron NP_858056.2
XM_005269028.4 457 Intron XP_005269085.1
XM_005269029.4 457 Intron XP_005269086.1
XM_005269032.3 457 Intron XP_005269089.1
XM_006719504.3 457 Intron XP_006719567.1
XM_006719505.3 457 Intron XP_006719568.1
XM_006719506.3 457 Intron XP_006719569.1
XM_006719507.3 457 Intron XP_006719570.1
XM_006719508.3 457 Intron XP_006719571.1
XM_006719509.3 457 Intron XP_006719572.1
XM_006719510.3 457 Intron XP_006719573.1
XM_006719512.3 457 Intron XP_006719575.1
XM_006719513.3 457 Intron XP_006719576.1
XM_006719514.3 457 Intron XP_006719577.1
XM_011538571.2 457 Intron XP_011536873.1
XM_017019651.1 457 Intron XP_016875140.1
XM_017019652.1 457 Intron XP_016875141.1
XM_017019653.1 457 Intron XP_016875142.1
XM_017019654.1 457 Intron XP_016875143.1
XM_017019655.1 457 Intron XP_016875144.1
XM_017019656.1 457 Intron XP_016875145.1
XM_017019657.1 457 Intron XP_016875146.1
XM_017019658.1 457 Intron XP_016875147.1
XM_017019659.1 457 Intron XP_016875148.1
XM_017019660.1 457 Intron XP_016875149.1
XM_017019661.1 457 Intron XP_016875150.1
XM_017019662.1 457 Intron XP_016875151.1
XM_017019663.1 457 Intron XP_016875152.1
XM_017019664.1 457 Intron XP_016875153.1
XM_017019665.1 457 Intron XP_016875154.1
Gene
FAM71C
Gene Name
family with sequence similarity 71 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153364.3 457 Missense Mutation ACG,ATG T11M NP_699195.1

View Full Product Details