Product Details

SNP ID
rs146726863
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6445132 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATCCCTGGTGGCTGTGCGTTCTG[A/G]GGACCCTGGTGGGGCTCTCAGCTAC
Phenotype
MIM: 186711 MIM: 607081
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD27 PubMed Links

Gene Details

Gene
CD27
Gene Name
CD27 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242.4 266 Missense Mutation AGG,GGG R13G NP_001233.1
XM_011521042.2 266 Intron XP_011519344.1
XM_017020232.1 266 Missense Mutation AGG,GGG R13G XP_016875721.1
XM_017020233.1 266 Intron XP_016875722.1
XM_017020234.1 266 Missense Mutation AGG,GGG R13G XP_016875723.1
Gene
CD27-AS1
Gene Name
CD27 antisense RNA 1
There are no transcripts associated with this gene.

Gene
TAPBPL
Gene Name
TAP binding protein like
There are no transcripts associated with this gene.

View Full Product Details