Product Details

SNP ID
rs147749635
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6774733 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCGAGTCCCTGTCCGGGAGTCCC[C/T]CCATCACCACTTAGCGGAAAGCTTC
Phenotype
MIM: 153337 MIM: 168440
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LAG3 PubMed Links

Gene Details

Gene
LAG3
Gene Name
lymphocyte activating 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002286.5 1020 Missense Mutation CCC,CTC P217L NP_002277.4
XM_011520956.1 1020 Missense Mutation CCC,CTC P217L XP_011519258.1
Gene
PTMS
Gene Name
parathymosin
There are no transcripts associated with this gene.

View Full Product Details