Product Details
- SNP ID
-
rs148085829
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:9754734 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGTTTGTTACATTAAACACCCTTC[G/T]GGGGAAGTAGATAGTAAATCCTGTT
- Phenotype
-
MIM: 107273
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CD69
PubMed Links
Gene Details
- Gene
- CD69
- Gene Name
- CD69 molecule
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001781.2 |
|
Intron |
|
|
NP_001772.1 |
View Full Product Details