Product Details

SNP ID
rs148322943
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:102958710 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCGCGGCCAACAAGAAGATGAGT[A/G]AGGTGGAGACACTGCGCTCGGCGGT
Phenotype
MIM: 100790
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASCL1 PubMed Links

Gene Details

Gene
ASCL1
Gene Name
achaete-scute family bHLH transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004316.3 1037 Missense Mutation AAG,GAG K156E NP_004307.2

View Full Product Details