Product Details
- SNP ID
-
rs149061964
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:120212562 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCGTGCTCGAAGAAGCTGCCGTTCA[C/T]GAATGGCGTGAAGCATTCCTGCCAG
- Phenotype
-
MIM: 602505
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PXN
PubMed Links
Gene Details
- Gene
- PXN
- Gene Name
- paxillin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001080855.2 |
2351 |
Missense Mutation |
ATG,GTG |
M510V |
NP_001074324.1 |
NM_001243756.1 |
2351 |
Missense Mutation |
ATG,GTG |
M524V |
NP_001230685.1 |
NM_002859.3 |
2351 |
Missense Mutation |
ATG,GTG |
M476V |
NP_002850.2 |
NM_025157.4 |
2351 |
Missense Mutation |
ATG,GTG |
M343V |
NP_079433.3 |
XM_005253917.3 |
2351 |
Missense Mutation |
ATG,GTG |
M322V |
XP_005253974.1 |
XM_006719532.1 |
2351 |
Missense Mutation |
ATG,GTG |
M1048V |
XP_006719595.1 |
XM_006719534.1 |
2351 |
Missense Mutation |
ATG,GTG |
M915V |
XP_006719597.1 |
XM_006719535.3 |
2351 |
Missense Mutation |
ATG,GTG |
M915V |
XP_006719598.1 |
XM_006719536.2 |
2351 |
Missense Mutation |
ATG,GTG |
M915V |
XP_006719599.1 |
XM_011538622.1 |
2351 |
Missense Mutation |
ATG,GTG |
M915V |
XP_011536924.1 |
XM_011538623.1 |
2351 |
Missense Mutation |
ATG,GTG |
M915V |
XP_011536925.1 |
XM_017019726.1 |
2351 |
Missense Mutation |
ATG,GTG |
M1054V |
XP_016875215.1 |
XM_017019727.1 |
2351 |
Missense Mutation |
ATG,GTG |
M1049V |
XP_016875216.1 |
XM_017019728.1 |
2351 |
Missense Mutation |
ATG,GTG |
M1046V |
XP_016875217.1 |
XM_017019729.1 |
2351 |
Missense Mutation |
ATG,GTG |
M1006V |
XP_016875218.1 |
XM_017019730.1 |
2351 |
UTR 3 |
|
|
XP_016875219.1 |
XM_017019731.1 |
2351 |
Missense Mutation |
ATG,GTG |
M915V |
XP_016875220.1 |
XM_017019732.1 |
2351 |
Intron |
|
|
XP_016875221.1 |
XM_017019733.1 |
2351 |
Missense Mutation |
ATG,GTG |
M859V |
XP_016875222.1 |
XM_017019734.1 |
2351 |
Missense Mutation |
ATG,GTG |
M807V |
XP_016875223.1 |
XM_017019735.1 |
2351 |
Missense Mutation |
ATG,GTG |
M777V |
XP_016875224.1 |
XM_017019736.1 |
2351 |
Missense Mutation |
ATG,GTG |
M667V |
XP_016875225.1 |
XM_017019737.1 |
2351 |
Missense Mutation |
ATG,GTG |
M619V |
XP_016875226.1 |
XM_017019738.1 |
2351 |
Missense Mutation |
ATG,GTG |
M564V |
XP_016875227.1 |
XM_017019739.1 |
2351 |
Missense Mutation |
ATG,GTG |
M530V |
XP_016875228.1 |
XM_017019740.1 |
2351 |
Missense Mutation |
ATG,GTG |
M516V |
XP_016875229.1 |
XM_017019741.1 |
2351 |
Missense Mutation |
ATG,GTG |
M508V |
XP_016875230.1 |
XM_017019742.1 |
2351 |
Missense Mutation |
ATG,GTG |
M482V |
XP_016875231.1 |
XM_017019743.1 |
2351 |
Missense Mutation |
ATG,GTG |
M343V |
XP_016875232.1 |
- Gene
- PXN-AS1
- Gene Name
- PXN antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details