Product Details

SNP ID
rs149279854
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56453626 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCCAGGGCAAGGGAGAAGCCAA[C/T]GGCCAGGGCCACGGAGCCCAGTTGG
Phenotype
MIM: 154050 MIM: 603887
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIP PubMed Links

Gene Details

Gene
MIP
Gene Name
major intrinsic protein of lens fiber
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012064.3 269 Missense Mutation ATT,GTT I164V NP_036196.1
XM_011538354.1 269 Missense Mutation ATT,GTT I69V XP_011536656.1
XM_017019306.1 269 Missense Mutation ATT,GTT I45V XP_016874795.1
Gene
TIMELESS
Gene Name
timeless circadian clock
There are no transcripts associated with this gene.

View Full Product Details