Product Details

SNP ID
rs149297262
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:51329800 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCCATGGACAGAATACTTGCCAT[C/T]CACCAAGCAATGGAGGGGGCCCCCA
Phenotype
MIM: 605936 MIM: 130120
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BIN2 PubMed Links

Gene Details

Gene
BIN2
Gene Name
bridging integrator 2
There are no transcripts associated with this gene.

Gene
CELA1
Gene Name
chymotrypsin like elastase family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001971.5 684 Missense Mutation AAT,GAT N215D NP_001962.3

View Full Product Details