Product Details
- SNP ID
-
rs149363709
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:57474148 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGTGACCACATGAATGTCATCCCA[C/T]TCAGTACTGTCCAAATCTAACCGAC
- Phenotype
-
MIM: 610576
MIM: 165220
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ARHGAP9
PubMed Links
Gene Details
- Gene
- ARHGAP9
- Gene Name
- Rho GTPase activating protein 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001080156.2 |
2287 |
Silent Mutation |
GAA,GAG |
E420E |
NP_001073625.1 |
NM_001080157.1 |
2287 |
Silent Mutation |
GAA,GAG |
E604E |
NP_001073626.1 |
NM_001319850.1 |
2287 |
Silent Mutation |
GAA,GAG |
E694E |
NP_001306779.1 |
NM_001319851.1 |
2287 |
Silent Mutation |
GAA,GAG |
E293E |
NP_001306780.1 |
NM_001319852.1 |
2287 |
Silent Mutation |
GAA,GAG |
E439E |
NP_001306781.1 |
NM_032496.3 |
2287 |
Silent Mutation |
GAA,GAG |
E604E |
NP_115885.2 |
XM_005269083.2 |
2287 |
Silent Mutation |
GAA,GAG |
E439E |
XP_005269140.1 |
XM_005269084.2 |
2287 |
Silent Mutation |
GAA,GAG |
E408E |
XP_005269141.1 |
XM_005269085.2 |
2287 |
Silent Mutation |
GAA,GAG |
E408E |
XP_005269142.1 |
XM_011538656.2 |
2287 |
Silent Mutation |
GAA,GAG |
E604E |
XP_011536958.1 |
XM_011538657.2 |
2287 |
Silent Mutation |
GAA,GAG |
E592E |
XP_011536959.1 |
XM_011538658.2 |
2287 |
Silent Mutation |
GAA,GAG |
E573E |
XP_011536960.1 |
XM_011538659.2 |
2287 |
Silent Mutation |
GAA,GAG |
E604E |
XP_011536961.1 |
XM_017019800.1 |
2287 |
Silent Mutation |
GAA,GAG |
E439E |
XP_016875289.1 |
XM_017019801.1 |
2287 |
Silent Mutation |
GAA,GAG |
E420E |
XP_016875290.1 |
XM_017019802.1 |
2287 |
Silent Mutation |
GAA,GAG |
E389E |
XP_016875291.1 |
XM_017019803.1 |
2287 |
Silent Mutation |
GAA,GAG |
E420E |
XP_016875292.1 |
XM_017019804.1 |
2287 |
Silent Mutation |
GAA,GAG |
E389E |
XP_016875293.1 |
- Gene
- GLI1
- Gene Name
- GLI family zinc finger 1
There are no transcripts associated with this gene.
View Full Product Details