Product Details

SNP ID
rs149847130
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48858097 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGTCGTTTGGAGAGGCTTCGGA[C/T]AGGGCTCTTCTGGGGCAGTGGGCTA
Phenotype
MIM: 612172 MIM: 609038
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DDX23 PubMed Links

Gene Details

Gene
DDX23
Gene Name
DEAD-box helicase 23
There are no transcripts associated with this gene.

Gene
RND1
Gene Name
Rho family GTPase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014470.3 701 Missense Mutation ATC,GTC I200V NP_055285.1

View Full Product Details