Product Details

SNP ID
rs150202472
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:69610268 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTCCACATCAATCACCTCCAGGA[A/T]GGGCATGTGAAGCAGCACAGTGGGA
Phenotype
MIM: 605139 MIM: 610846
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CCT2 PubMed Links

Gene Details

Gene
CCT2
Gene Name
chaperonin containing TCP1 subunit 2
There are no transcripts associated with this gene.

Gene
LRRC10
Gene Name
leucine rich repeat containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_201550.3 895 Missense Mutation ATC,TTC I191F NP_963844.2

View Full Product Details