Product Details

SNP ID
rs150228563
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:99648642 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATCTTCAGCTGTGTCCTCCATC[C/G]GATGCAAGTGAAGACCTTTTTGTTC
Phenotype
MIM: 607815
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ANKS1B PubMed Links

Gene Details

Gene
ANKS1B
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204065.1 893 Intron NP_001190994.1
NM_001204066.1 893 Intron NP_001190995.1
NM_001204067.1 893 Intron NP_001190996.1
NM_001204068.1 893 Intron NP_001190997.1
NM_001204069.1 893 Intron NP_001190998.1
NM_001204070.1 893 Intron NP_001190999.1
NM_001204079.1 893 Intron NP_001191008.1
NM_001204080.1 893 Intron NP_001191009.1
NM_001204081.1 893 Intron NP_001191010.1
NM_020140.3 893 Intron NP_064525.1
NM_152788.4 893 Intron NP_690001.3
NM_181670.3 893 Intron NP_858056.2
XM_005269028.4 893 Intron XP_005269085.1
XM_005269029.4 893 Intron XP_005269086.1
XM_005269032.3 893 Intron XP_005269089.1
XM_006719504.3 893 Intron XP_006719567.1
XM_006719505.3 893 Intron XP_006719568.1
XM_006719506.3 893 Intron XP_006719569.1
XM_006719507.3 893 Intron XP_006719570.1
XM_006719508.3 893 Intron XP_006719571.1
XM_006719509.3 893 Intron XP_006719572.1
XM_006719510.3 893 Intron XP_006719573.1
XM_006719512.3 893 Intron XP_006719575.1
XM_006719513.3 893 Intron XP_006719576.1
XM_006719514.3 893 Intron XP_006719577.1
XM_011538571.2 893 Intron XP_011536873.1
XM_017019651.1 893 Intron XP_016875140.1
XM_017019652.1 893 Intron XP_016875141.1
XM_017019653.1 893 Intron XP_016875142.1
XM_017019654.1 893 Intron XP_016875143.1
XM_017019655.1 893 Intron XP_016875144.1
XM_017019656.1 893 Intron XP_016875145.1
XM_017019657.1 893 Intron XP_016875146.1
XM_017019658.1 893 Intron XP_016875147.1
XM_017019659.1 893 Intron XP_016875148.1
XM_017019660.1 893 Intron XP_016875149.1
XM_017019661.1 893 Intron XP_016875150.1
XM_017019662.1 893 Intron XP_016875151.1
XM_017019663.1 893 Intron XP_016875152.1
XM_017019664.1 893 Intron XP_016875153.1
XM_017019665.1 893 Intron XP_016875154.1
Gene
FAM71C
Gene Name
family with sequence similarity 71 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153364.3 893 Silent Mutation TCC,TCG S156S NP_699195.1

View Full Product Details