Product Details

SNP ID
rs150331875
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:123771849 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGGCCACCCTCAGATTCAGTGAT[G/T]GGCTGGAAGGACTCACAGGACCCAG
Phenotype
MIM: 605884
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DNAH10 PubMed Links

Gene Details

Gene
DNAH10
Gene Name
dynein axonemal heavy chain 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207437.3 Intron NP_997320.2
XM_005253555.3 Intron XP_005253612.1
XM_011538014.2 Intron XP_011536316.1
XM_011538015.2 Intron XP_011536317.1
XM_011538016.2 Intron XP_011536318.1
XM_011538017.2 Intron XP_011536319.1
XM_011538019.2 Intron XP_011536321.1
XM_011538020.2 Intron XP_011536322.1
XM_017018960.1 Intron XP_016874449.1
XM_017018961.1 Intron XP_016874450.1
XM_017018962.1 Intron XP_016874451.1

View Full Product Details