Product Details

SNP ID
rs150735821
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:112405964 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGGATTTTTACATTGCTGATATC[A/G]ATTTTGGTTGAAGTGGCAATGACAA
Phenotype
MIM: 603703
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPL6 PubMed Links

Gene Details

Gene
RPL6
Gene Name
ribosomal protein L6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000970.4 882 Intron NP_000961.2
NM_001024662.2 882 Intron NP_001019833.1
NM_001320137.1 882 Intron NP_001307066.1
NM_001320138.1 882 Silent Mutation ATC,ATT I201I NP_001307067.1
NM_001320139.1 882 Intron NP_001307068.1
NM_001320140.1 882 Silent Mutation ATC,ATT I201I NP_001307069.1
NM_001320141.1 882 Silent Mutation ATC,ATT I201I NP_001307070.1
NM_001320142.1 882 Intron NP_001307071.1
XM_017019781.1 882 Silent Mutation ATC,ATT I201I XP_016875270.1
XM_017019782.1 882 Intron XP_016875271.1

View Full Product Details