Product Details

SNP ID
rs151006044
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:71689598 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTTTATAATTTTTCAGGTAACTT[A/T]CGACCTATTTCTCCGTGGCGTTGGC
Phenotype
MIM: 612531
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
THAP2 PubMed Links

Gene Details

Gene
THAP2
Gene Name
THAP domain containing 2
There are no transcripts associated with this gene.

Gene
TMEM19
Gene Name
transmembrane protein 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018279.3 721 Missense Mutation TTA,TTT L46F NP_060749.2

View Full Product Details