Product Details
- SNP ID
-
rs151011578
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:106066903 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGCTGCTGTCTGCCAGCCGGTTCC[A/G]GTACCGCTTCAGGTACTGGGGCCGG
- Phenotype
-
MIM: 608130
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NUAK1
PubMed Links
Gene Details
- Gene
- NUAK1
- Gene Name
- NUAK family kinase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014840.2 |
3265 |
Missense Mutation |
CGG,TGG |
R629W |
NP_055655.1 |
View Full Product Details