Product Details

SNP ID
rs151038965
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:12474027 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTAAAGAATGAAGGCTGGCACTAA[A/C]CTGTTAGCTAATCATCCGGTGTTCC
Phenotype
MIM: 616598 MIM: 607175
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BORCS5 PubMed Links

Gene Details

Gene
BORCS5
Gene Name
BLOC-1 related complex subunit 5
There are no transcripts associated with this gene.

Gene
DUSP16
Gene Name
dual specificity phosphatase 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030640.2 5567 UTR 3 NP_085143.1
XM_005253488.2 5567 Intron XP_005253545.1
XM_006719155.2 5567 Intron XP_006719218.1
XM_011520856.1 5567 Intron XP_011519158.1
XM_011520857.1 5567 Intron XP_011519159.1
XM_017019988.1 5567 Intron XP_016875477.1
Gene
LOC105369662
Gene Name
uncharacterized LOC105369662
There are no transcripts associated with this gene.

View Full Product Details