Product Details

SNP ID
rs151102289
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56120745 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAGCGTTGCCGATATCGCCACCC[A/G]GACATGAGCGAGGTGTCCAACTTGG
Phenotype
MIM: 616670 MIM: 602145 MIM: 613315
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ESYT1 PubMed Links

Gene Details

Gene
ESYT1
Gene Name
extended synaptotagmin 1
There are no transcripts associated with this gene.

Gene
PA2G4
Gene Name
proliferation-associated 2G4
There are no transcripts associated with this gene.

Gene
RPL41
Gene Name
ribosomal protein L41
There are no transcripts associated with this gene.

Gene
ZC3H10
Gene Name
zinc finger CCCH-type containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303124.1 507 Silent Mutation CCA,CCG P61P NP_001290053.1
NM_001303125.1 507 Silent Mutation CCA,CCG P61P NP_001290054.1
NM_032786.2 507 Silent Mutation CCA,CCG P61P NP_116175.1

View Full Product Details