Product Details

SNP ID
rs137952665
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:99201551 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGATGTTGCTGTTTTCCTGGCGTG[C/T]TAGCGGTGGTCAGCAGGTAGGGGGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UBAC2 PubMed Links

Gene Details

Gene
UBAC2
Gene Name
UBA domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144072.1 206 Intron NP_001137544.1
NM_177967.3 206 Missense Mutation GCT,GTT A47V NP_808882.1
XM_006719948.3 206 UTR 5 XP_006720011.1
XM_011521082.2 206 Intron XP_011519384.1
XM_011521083.2 206 Missense Mutation GCT,GTT A30V XP_011519385.1
XM_011521084.2 206 Intron XP_011519386.1
XM_017020553.1 206 Intron XP_016876042.1
Gene
UBAC2-AS1
Gene Name
UBAC2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details