Product Details

SNP ID
rs138507433
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:52653071 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTTTCCAGAGCTTCTCGGATGCC[C/T]TAATCGACGAGGACCCCCAGGCGGC
Phenotype
MIM: 604098
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HNRNPA1L2 PubMed Links

Gene Details

Gene
HNRNPA1L2
Gene Name
heterogeneous nuclear ribonucleoprotein A1-like 2
There are no transcripts associated with this gene.

Gene
SUGT1
Gene Name
SGT1 homolog, MIS12 kinetochore complex assembly cochaperone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130912.2 294 Silent Mutation CTA,TTA L22L NP_001124384.1
NM_001320831.1 294 UTR 5 NP_001307760.1
NM_006704.4 294 Silent Mutation CTA,TTA L22L NP_006695.1
XM_011534891.2 294 UTR 5 XP_011533193.1
XM_011534893.2 294 UTR 5 XP_011533195.1
XM_017020365.1 294 Silent Mutation CTA,TTA L22L XP_016875854.1
XM_017020366.1 294 UTR 5 XP_016875855.1
XM_017020367.1 294 Intron XP_016875856.1
XM_017020368.1 294 UTR 5 XP_016875857.1

View Full Product Details