Product Details
- SNP ID
-
rs138515715
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:27793912 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATTGCCCATGTCTCCGTCCTCCTC[A/G]GGGAAGGACGACCGGGATCTTACGG
- Phenotype
-
MIM: 616542
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GSX1
PubMed Links
Gene Details
- Gene
- GSX1
- Gene Name
- GS homeobox 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_145657.2 |
807 |
Silent Mutation |
TCA,TCG |
S253S |
NP_663632.1 |
View Full Product Details