Product Details
- SNP ID
-
rs139262236
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:106544465 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAATTACATCTACATTTTCTGTCTT[C/T]CATTTTCAATGAAAATAAATGAACA
- Phenotype
-
MIM: 614046
MIM: 600527
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ARGLU1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs150852690] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ARGLU1
- Gene Name
- arginine and glutamate rich 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018011.3 |
|
Intron |
|
|
NP_060481.3 |
- Gene
- EFNB2
- Gene Name
- ephrin B2
There are no transcripts associated with this gene.
View Full Product Details